Assay Details
| Gene Symbol: ABCG33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336645.1 | NP_001325069.1 | 1 - 2 | 521 | 131 | |
| NM_001336646.1 | NP_001325068.1 | 1 - 2 | 455 | 131 | ||
| NM_001336647.1 | NP_001325070.1 | 1 - 2 | 455 | 131 | ||
| NM_129284.2 | NP_181265.1 | 1 - 2 | 521 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001336647.1 transcripts.
Set Membership: |
|

ABCG33
Genomic Map