Assay Details
| Gene Symbol: AT2G37520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129307.5 | NP_181288.4 | 2 - 3 | 1188 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G37520| Gene Symbol: AT2G37520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129307.5 | NP_181288.4 | 2 - 3 | 1188 | 120 | |
Set Membership: |
|