Assay Details
| Gene Symbol: SLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129312.4 | NP_565864.1 | 1 | 827 | 104 | |
| GenBank mRNA | AF213400.1 | - | 1 | 326 | 104 | |
| AY054667.1 | - | 1 | 572 | 104 | ||
| AY081508.1 | - | 1 | 326 | 104 | ||
More Information
Set Membership: |
|

SLT1
Genomic Map