Assay Details
| Gene Symbol: AT2G38000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129357.4 | NP_181336.1 | 5 - 6 | 865 | 83 | |
| GenBank mRNA | AY080880.1 | - | 5 - 6 | 845 | 83 | |
| AY114047.1 | - | 5 - 6 | 750 | 83 | ||
More Information
Set Membership: |
|

AT2G38000
Genomic Map