Assay Details
| Gene Symbol: VCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129359.4 | NP_565879.1 | 15 - 16 | 2530 | 67 | |
| GenBank mRNA | AF359240.1 | - | 14 - 15 | 2437 | 67 | |
| AY048248.1 | - | 15 - 16 | 2504 | 67 | ||
| AY139799.1 | - | 14 - 15 | 2421 | 67 | ||
More Information
Set Membership: |
|

VCL1
Genomic Map