Assay Details
| Gene Symbol: VCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084551.1 | NP_001078020.1 | 5 - 6 | 1492 | 104 | |
| NM_129359.4 | NP_565879.1 | 5 - 6 | 1493 | 104 | ||
| GenBank mRNA | AF359240.1 | - | 4 - 5 | 1400 | 104 | |
| AY048248.1 | - | 5 - 6 | 1467 | 104 | ||
| AY139799.1 | - | 4 - 5 | 1384 | 104 | ||
More Information
Set Membership: |
|

VCL1
Genomic Map