Assay Details
| Gene Symbol: VCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084551.1 | NP_001078020.1 | 7 - 8 | 1669 | 87 | |
| NM_129359.4 | NP_565879.1 | 7 - 8 | 1670 | 87 | ||
| GenBank mRNA | AF359240.1 | - | 6 - 7 | 1577 | 87 | |
| AY048248.1 | - | 7 - 8 | 1644 | 87 | ||
| AY139799.1 | - | 6 - 7 | 1561 | 87 | ||
More Information
Set Membership: |
|

VCL1
Genomic Map