Assay Details
| Gene Symbol: GPAT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129367.4 | NP_181346.1 | 1 - 2 | 1027 | 120 | |
| GenBank mRNA | AY062721.1 | - | 1 - 2 | 769 | 120 | |
| AY114657.1 | - | 1 - 2 | 639 | 120 | ||
More Information
Set Membership: |
|

GPAT6
Genomic Map