Assay Details
| Gene Symbol: AUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336689.1 | NP_001324970.1 | 2 - 3 | 471 | 128 | |
| NM_129368.3 | NP_565882.1 | 2 - 3 | 588 | 128 | ||
| GenBank mRNA | AY054488.1 | - | 2 - 3 | 471 | 128 | |
| AY087092.1 | - | 2 - 3 | 466 | 128 | ||
| AY093300.1 | - | 1 - 2 | 182 | 128 | ||
More Information
Set Membership: |
|

AUX1
Genomic Map