Assay Details
| Gene Symbol: AUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336689.1 | NP_001324970.1 | 6 - 7 | 1040 | 147 | |
| NM_129368.3 | NP_565882.1 | 6 - 7 | 1157 | 147 | ||
| GenBank mRNA | AY054488.1 | - | 6 - 7 | 1040 | 147 | |
| AY087092.1 | - | 6 - 7 | 1035 | 147 | ||
| AY093300.1 | - | 5 - 6 | 751 | 147 | ||
More Information
Set Membership: |
|

AUX1
Genomic Map