Assay Details
| Gene Symbol: AUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129368.3 | NP_565882.1 | 8 - 9 | 1634 | 78 | |
| GenBank mRNA | AY054488.1 | - | 8 - 9 | 1517 | 78 | |
| AY087092.1 | - | 8 - 9 | 1512 | 78 | ||
| AY093300.1 | - | 7 - 8 | 1228 | 78 | ||
More Information
Set Membership: |
|

AUX1
Genomic Map