Assay Details
| Gene Symbol: AT2G38180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129374.4 | NP_565883.1 | 4 - 5 | 665 | 78 | |
| GenBank mRNA | BT021087.1 | - | 4 - 5 | 419 | 78 | |
| BT021116.1 | - | 4 - 5 | 412 | 78 | ||
More Information
Set Membership: |
|

AT2G38180
Genomic Map