Assay Details
| Gene Symbol: AMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129385.5 | NP_181363.1 | 3 - 4 | 1119 | 79 | |
| NM_201905.1 | NP_973634.1 | 2 - 3 | 1060 | 79 | ||
| GenBank mRNA | AK226212.1 | - | 3 - 4 | 919 | 79 | |
| BT025321.1 | - | 3 - 4 | 889 | 79 | ||
More Information
Set Membership: |
|

AMT2
Genomic Map