Assay Details
| Gene Symbol: AT2G38300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129386.3 | NP_181364.2 | 1 - 2 | 570 | 89 | |
| GenBank mRNA | AK117689.1 | - | 1 - 2 | 547 | 89 | |
| BT004973.1 | - | 1 - 2 | 234 | 89 | ||
More Information
Set Membership: |
|

AT2G38300
Genomic Map