Assay Details
| Gene Symbol: AGT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202772.1 | NP_001189701.1 | 2 - 3 | 874 | 67 | |
| NM_129396.4 | NP_181374.1 | 2 - 3 | 920 | 67 | ||
| GenBank mRNA | AF166352.1 | - | 2 - 3 | 846 | 67 | |
| AY085348.1 | - | 2 - 3 | 874 | 67 | ||
| BT001918.1 | - | 2 - 3 | 786 | 67 | ||
More Information
Set Membership: |
|

AGT3
Genomic Map