Assay Details
| Gene Symbol: AGT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129396.4 | NP_181374.1 | 6 - 7 | 1525 | 63 | |
| GenBank mRNA | AF166352.1 | - | 6 - 7 | 1451 | 63 | |
| AY085348.1 | - | 6 - 7 | 1479 | 63 | ||
| BT001918.1 | - | 6 - 7 | 1391 | 63 | ||
More Information
Set Membership: |
|

AGT3
Genomic Map