Assay Details
| Gene Symbol: ANNAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129433.5 | NP_181410.1 | 1 - 2 | 218 | 79 | |
| GenBank mRNA | AK227695.1 | - | 1 - 2 | 145 | 79 | |
| AY087221.1 | - | 1 - 2 | 148 | 79 | ||
| BT006420.1 | - | 1 - 2 | 76 | 79 | ||
More Information
Set Membership: |
|

ANNAT3
Genomic Map