Assay Details
| Gene Symbol: ANNAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129433.5 | NP_181410.1 | 4 - 5 | 804 | 86 | |
| GenBank mRNA | AK227695.1 | - | 4 - 5 | 731 | 86 | |
| AY087221.1 | - | 4 - 5 | 734 | 86 | ||
| BT006420.1 | - | 4 - 5 | 662 | 86 | ||
More Information
Set Membership: |
|

ANNAT3
Genomic Map