Assay Details
| Gene Symbol: PIP2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129458.3 | NP_181434.1 | 2 - 3 | 878 | 70 | |
| GenBank mRNA | AY045690.1 | - | 2 - 3 | 678 | 70 | |
| AY054142.1 | - | 2 - 3 | 594 | 70 | ||
| AY057559.1 | - | 2 - 3 | 676 | 70 | ||
More Information
Set Membership: |
|

PIP2E
Genomic Map