Assay Details
| Gene Symbol: AT2G39040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129461.3 | NP_181437.1 | 2 - 3 | 535 | 67 | |
| GenBank mRNA | BT026452.1 | - | 2 - 3 | 463 | 67 | |
More Information
Set Membership: |
|

AT2G39040
Genomic Map