Assay Details
| Gene Symbol: AT2G39040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129461.3 | NP_181437.1 | 3 - 4 | 710 | 70 | |
| GenBank mRNA | BT026452.1 | - | 3 - 4 | 638 | 70 | |
More Information
Set Membership: |
|

AT2G39040
Genomic Map