Assay Details
| Gene Symbol: SVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336760.1 | NP_001323702.1 | 5 - 6 | 969 | 104 | |
| NM_129471.3 | NP_181447.2 | 5 - 6 | 1023 | 104 | ||
| GenBank mRNA | AK117682.1 | - | 5 - 6 | 1008 | 104 | |
| AY088622.1 | - | 5 - 6 | 956 | 104 | ||
More Information
Set Membership: |
|

SVR1
Genomic Map