Assay Details
| Gene Symbol: SVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129471.3 | NP_181447.2 | 8 - 9 | 1282 | 66 | |
| GenBank mRNA | AK117682.1 | - | 8 - 9 | 1267 | 66 | |
| AY088622.1 | - | 8 - 9 | 1215 | 66 | ||
More Information
Set Membership: |
|

SVR1
Genomic Map