Assay Details
| Gene Symbol: PLP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129480.3 | NP_181455.1 | 1 - 2 | 1696 | 67 | |
| GenBank mRNA | AY062648.1 | - | 1 - 2 | 1548 | 67 | |
| BT000763.1 | - | 1 - 2 | 1310 | 67 | ||
| BT008869.1 | - | 1 - 2 | 1191 | 67 | ||
More Information
Set Membership: |
|

PLP6
Genomic Map