Assay Details
| Gene Symbol: PGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129486.5 | NP_181461.1 | 2 - 3 | 605 | 57 | |
| GenBank mRNA | AB048535.1 | - | 2 - 3 | 481 | 57 | |
| AB058617.1 | - | 2 - 3 | 481 | 57 | ||
| BT024857.1 | - | 2 - 3 | 466 | 57 | ||
More Information
Set Membership: |
|

PGP1
Genomic Map