Assay Details
| Gene Symbol: SAC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129491.5 | NP_181466.2 | 7 - 8 | 2127 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAC3A| Gene Symbol: SAC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129491.5 | NP_181466.2 | 7 - 8 | 2127 | 119 | |
Set Membership: |
|