Assay Details
| Gene Symbol: MTP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129503.4 | NP_181477.1 | 3 - 4 | 847 | 100 | |
| GenBank mRNA | AY136449.1 | - | 3 - 4 | 609 | 100 | |
| BT008477.1 | - | 3 - 4 | 522 | 100 | ||
More Information
Set Membership: |
|

MTP11
Genomic Map