Assay Details
Gene Symbol: MTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129503.4 | NP_181477.1 | 5 - 6 | 1161 | 113 | |
GenBank mRNA | AY136449.1 | - | 5 - 6 | 923 | 113 | |
BT008477.1 | - | 5 - 6 | 836 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
