Assay Details
| Gene Symbol: AT2G39530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129511.3 | NP_181485.2 | 1 - 2 | 306 | 69 | |
| GenBank mRNA | AK118915.1 | - | 1 - 2 | 253 | 69 | |
| BT005644.1 | - | 1 - 2 | 183 | 69 | ||
More Information
Set Membership: |
|

AT2G39530
Genomic Map