Assay Details
| Gene Symbol: BIK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129522.5 | NP_181496.1 | 3 - 4 | 912 | 98 | |
| GenBank mRNA | AF325086.1 | - | 3 - 4 | 496 | 98 | |
| AY062493.1 | - | 3 - 4 | 714 | 98 | ||
| AY065029.1 | - | 3 - 4 | 714 | 98 | ||
| AY093278.1 | - | 3 - 4 | 496 | 98 | ||
| AY093997.1 | - | 3 - 4 | 496 | 98 | ||
More Information
Set Membership: |
|

BIK1
Genomic Map