Assay Details
| Gene Symbol: P5CS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202785.1 | NP_001189714.1 | 15 - 16 | 1834 | 77 | |
| NM_001202786.1 | NP_001189715.1 | 15 - 16 | 1825 | 77 | ||
| NM_129539.2 | NP_181510.1 | 15 - 16 | 2092 | 77 | ||
| NM_201912.1 | NP_973641.1 | 14 - 15 | 1728 | 77 | ||
| GenBank mRNA | AF424633.1 | - | 15 - 16 | 1831 | 77 | |
| AK316756.1 | - | 14 - 15 | 1728 | 77 | ||
| AY113046.1 | - | 15 - 16 | 1712 | 77 | ||
| AY150430.1 | - | 15 - 16 | 1712 | 77 | ||
| X86777.1 | - | 1784 | 77 | |||
| X87330.1 | - | 1810 | 77 | |||
More Information
Set Membership: |
|

P5CS1
Genomic Map