Assay Details
| Gene Symbol: P5CS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202785.1 | NP_001189714.1 | 16 - 17 | 1989 | 107 | |
| NM_001202786.1 | NP_001189715.1 | 16 - 17 | 1980 | 107 | ||
| NM_129539.2 | NP_181510.1 | 16 - 17 | 2247 | 107 | ||
| NM_201912.1 | NP_973641.1 | 15 - 16 | 1883 | 107 | ||
| GenBank mRNA | AF424633.1 | - | 16 - 17 | 1986 | 107 | |
| AK316756.1 | - | 15 - 16 | 1883 | 107 | ||
| AY113046.1 | - | 16 - 17 | 1867 | 107 | ||
| AY150430.1 | - | 16 - 17 | 1867 | 107 | ||
| X86777.1 | - | 1939 | 107 | |||
| X87330.1 | - | 1965 | 107 | |||
More Information
Set Membership: |
|

P5CS1
Genomic Map