Assay Details
| Gene Symbol: P5CS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202785.1 | NP_001189714.1 | 3 - 4 | 359 | 73 | |
| NM_001202786.1 | NP_001189715.1 | 3 - 4 | 359 | 73 | ||
| NM_129539.2 | NP_181510.1 | 3 - 4 | 617 | 73 | ||
| GenBank mRNA | AF424633.1 | - | 3 - 4 | 356 | 73 | |
| AY113046.1 | - | 3 - 4 | 237 | 73 | ||
| AY150430.1 | - | 3 - 4 | 237 | 73 | ||
| X86777.1 | - | 309 | 73 | |||
| X87330.1 | - | 335 | 73 | |||
More Information
Set Membership: |
|

P5CS1
Genomic Map