Assay Details
| Gene Symbol: P5CS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202785.1 | NP_001189714.1 | 6 - 7 | 902 | 86 | |
| NM_001202786.1 | NP_001189715.1 | 6 - 7 | 902 | 86 | ||
| NM_129539.2 | NP_181510.1 | 6 - 7 | 1160 | 86 | ||
| NM_201912.1 | NP_973641.1 | 5 - 6 | 796 | 86 | ||
| GenBank mRNA | AF424633.1 | - | 6 - 7 | 899 | 86 | |
| AK316756.1 | - | 5 - 6 | 796 | 86 | ||
| AY113046.1 | - | 6 - 7 | 780 | 86 | ||
| AY150430.1 | - | 6 - 7 | 780 | 86 | ||
| X86777.1 | - | 852 | 86 | |||
| X87330.1 | - | 878 | 86 | |||
More Information
Set Membership: |
|

P5CS1
Genomic Map