Assay Details
| Gene Symbol: HOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336798.1 | NP_001324757.1 | 1 - 2 | 238 | 104 | |
| NM_129540.5 | NP_181511.3 | 1 - 2 | 255 | 104 | ||
| GenBank mRNA | BT004255.1 | - | 1 - 2 | 196 | 104 | |
| BT005517.1 | - | 1 - 2 | 91 | 104 | ||
More Information
Set Membership: |
|

HOS1
Genomic Map