Assay Details
| Gene Symbol: HOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336798.1 | NP_001324757.1 | 10 - 11 | 2683 | 96 | |
| NM_129540.5 | NP_181511.3 | 9 - 10 | 2779 | 96 | ||
| GenBank mRNA | BT004255.1 | - | 9 - 10 | 2720 | 96 | |
| BT005517.1 | - | 9 - 10 | 2615 | 96 | ||
More Information
Set Membership: |
|

HOS1
Genomic Map