Assay Details
| Gene Symbol: DAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336799.1 | NP_001324813.1 | 3 - 4 | 848 | 92 | |
| NM_001336800.1 | NP_001324811.1 | 3 - 4 | 821 | 92 | ||
| NM_001336801.1 | NP_001324812.1 | 3 - 4 | 557 | 92 | ||
| NM_129542.5 | NP_181513.3 | 3 - 4 | 735 | 92 | ||
| GenBank mRNA | AK227895.1 | - | 3 - 4 | 476 | 92 | |
More Information
Set Membership: |
|

DAR2
Genomic Map