Assay Details
| Gene Symbol: DAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336799.1 | NP_001324813.1 | 9 - 10 | 1577 | 84 | |
| NM_001336800.1 | NP_001324811.1 | 9 - 10 | 1550 | 84 | ||
| NM_001336801.1 | NP_001324812.1 | 9 - 10 | 1286 | 84 | ||
| NM_129542.5 | NP_181513.3 | 9 - 10 | 1464 | 84 | ||
More Information
Set Membership: |
|

DAR2
Genomic Map