Assay Details
| Gene Symbol: AT2G39910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129549.4 | NP_565917.2 | 6 - 7 | 1121 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G39910| Gene Symbol: AT2G39910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129549.4 | NP_565917.2 | 6 - 7 | 1121 | 81 | |
Set Membership: |
|