Assay Details
| Gene Symbol: EIF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129557.4 | NP_181528.1 | 4 - 5 | 1053 | 145 | |
| GenBank mRNA | AY045824.1 | - | 4 - 5 | 738 | 145 | |
| AY091363.1 | - | 4 - 5 | 673 | 145 | ||
| U54561.2 | - | 4 - 5 | 726 | 145 | ||
More Information
Set Membership: |
|

EIF2
Genomic Map