Assay Details
| Gene Symbol: LEW3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129577.5 | NP_181548.2 | 1 - 2 | 362 | 113 | |
| GenBank mRNA | KJ138857.1 | - | 1 - 2 | 303 | 113 | |
More Information
Set Membership: |
|

LEW3
Genomic Map