Assay Details
| Gene Symbol: FER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129588.5 | NP_181559.1 | 7 - 8 | 823 | 113 | |
| GenBank mRNA | AJ312191.1 | - | 7 - 8 | 785 | 113 | |
| AY062795.1 | - | 7 - 8 | 809 | 113 | ||
| AY081615.1 | - | 7 - 8 | 743 | 113 | ||
More Information
Set Membership: |
|

FER4
Genomic Map