Assay Details
| Gene Symbol: HEME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129611.5 | NP_181581.1 | 3 - 4 | 508 | 130 | |
| GenBank mRNA | AY142634.1 | - | 3 - 4 | 352 | 130 | |
More Information
Set Membership: |
|

HEME2
Genomic Map