Assay Details
| Gene Symbol: GLY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336858.1 | NP_001324724.1 | 1 - 2 | 381 | 72 | |
| NM_129631.3 | NP_565939.1 | 1 - 2 | 381 | 72 | ||
| GenBank mRNA | AY050968.1 | - | 1 - 2 | 344 | 72 | |
| AY114078.1 | - | 1 - 2 | 266 | 72 | ||
More Information
Set Membership: |
|

GLY1
Genomic Map