Assay Details
| Gene Symbol: GLY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336857.1 | NP_001324723.1 | 3 - 4 | 899 | 86 | |
| NM_001336858.1 | NP_001324724.1 | 4 - 5 | 825 | 86 | ||
| NM_129631.3 | NP_565939.1 | 4 - 5 | 825 | 86 | ||
| GenBank mRNA | AY050968.1 | - | 4 - 5 | 788 | 86 | |
| AY114078.1 | - | 4 - 5 | 710 | 86 | ||
More Information
Set Membership: |
|

GLY1
Genomic Map