Assay Details
| Gene Symbol: CTEXP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129635.5 | NP_181605.2 | 17 - 18 | 1916 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTEXP| Gene Symbol: CTEXP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129635.5 | NP_181605.2 | 17 - 18 | 1916 | 85 | |
Set Membership: |
|