Assay Details
| Gene Symbol: DPE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129647.4 | NP_181616.3 | 2 - 3 | 623 | 95 | |
| GenBank mRNA | AY081315.1 | - | 2 - 3 | 414 | 95 | |
| BT010364.1 | - | 1 - 2 | 365 | 95 | ||
More Information
Set Membership: |
|

DPE2
Genomic Map