Assay Details
| Gene Symbol: DPE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129647.4 | NP_181616.3 | 6 - 7 | 1045 | 79 | |
| GenBank mRNA | AY081315.1 | - | 6 - 7 | 836 | 79 | |
| BT010364.1 | - | 5 - 6 | 787 | 79 | ||
More Information
Set Membership: |
|

DPE2
Genomic Map