Assay Details
| Gene Symbol: DPE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129647.4 | NP_181616.3 | 8 - 9 | 1323 | 70 | |
| GenBank mRNA | AY081315.1 | - | 8 - 9 | 1114 | 70 | |
| BT010364.1 | - | 7 - 8 | 1065 | 70 | ||
More Information
Set Membership: |
|

DPE2
Genomic Map