Assay Details
| Gene Symbol: GLU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129687.5 | NP_181655.1 | 14 - 15 | 2851 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLU2| Gene Symbol: GLU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129687.5 | NP_181655.1 | 14 - 15 | 2851 | 148 | |
Set Membership: |
|